missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse CCL17/TARC Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 20295 |
Molecular Weight (g/mol) | 7.9 kDa |
Name | CCL17/TARC |
Quantity | 25μg |
Description
The Recombinant Mouse CCL17/TARC Protein is derived from E. coli.Specifications
Unconjugated | |
7.9 kDa | |
25μg | |
E. coli-derived mouse CCL17/TARC protein Ala24-Pro93 |
20295 | |
CCL17/TARC | |
Yes |