missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CCL11/Eotaxin Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 420-ME-100/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Código de producto. 13434364

  • £607.11 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Descripción

Descripción

The Recombinant Mouse CCL11/Eotaxin Protein is derived from E. coli. The Recombinant Mouse CCL11/Eotaxin Protein has been validated for the following applications: Bioactivity.
Especificaciones

Especificaciones

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
8.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
20292
CCL11/Eotaxin
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documentos