missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse BMP-7 Recombinant Protein

Brand:  R&D Systems™ 5666-BP-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464024

  • £429.02 / 10µg


Estimated Shipment
08-01-2019
Add to basket

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser292-His430
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 12162
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
15.6 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Unlabeled
12162
BMP-7
10μg
Disulfide-linked homodimer
Yes
Videos