missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse BMP-6 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 6325-BM-020

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445764

  • £588.00 / 20µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse BMP-6 Protein is derived from CHO. The Recombinant Mouse BMP-6 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
17-21 kDa, reducing conditions
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Mouse
Disulfide-linked homodimer
Yes
12161
BMP-6
20μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents