missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse BMP-6 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 12161 |
---|---|
Molecular Weight (g/mol) | 17-21 kDa, reducing conditions |
Name | BMP-6 |
Quantity | 20μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse BMP-6 protein Ser372-His510 |
Description
The Recombinant Mouse BMP-6 Protein is derived from CHO. The Recombinant Mouse BMP-6 Protein has been validated for the following applications: Bioactivity.Specifications
12161 | |
BMP-6 | |
Chinese Hamster Ovary cell line,CHO-derived mouse BMP-6 protein Ser372-His510 | |
Disulfide-linked homodimer | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
17-21 kDa, reducing conditions | |
20μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, VGR1, VGR-1 | |
Recombinant |