missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Betacellulin/BTC Recombinant Protein

Brand:  R&D Systems™ 1025-CE-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488323

  • £331.00 / 25µg


Estimated Shipment
02-07-2018
Add to basket

Description

Description

Source: E. coli-derived, Asp32-Gln118
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 12223
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
9.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
Yes
12223
Betacellulin/BTC
25μg
Unlabeled
Videos