missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Artemin Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 11876 |
Molecular Weight (g/mol) | 12 kDa (monomer) |
Name | Artemin |
Quantity | 25μg |
Description
The Recombinant Mouse Artemin Protein is derived from E. coli. The Recombinant Mouse Artemin Protein has been validated for the following applications: Bioactivity, Binding Activity.Specifications
Unconjugated | |
12 kDa (monomer) | |
25μg | |
E. coli-derived mouse Artemin protein Ala112-Gly224 |
11876 | |
Artemin | |
Yes |