missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse 4-1BB Ligand/TNFSF9 Recombinant Protein

Brand:  R&D Systems™ 1246-4L-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488963

  • £340.57 / 25µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Arg104-Glu309, with a N-terminal Met and 10-His tag + 16 aa linker
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21950
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
25.7 kDa
25μg
Mouse
21950
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
4-1BB Ligand/TNFSF9
Yes
Videos