missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Wnt-3a Recombinant Protein

Brand:  R&D Systems™ 5036-WN-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465254

  • £252.13 / 10µg


Estimated Shipment
22-10-2018
Add to basket

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser19-Lys352
Purity: 75%, by SDS-PAGE with silver staining.
Molecular Weight: 37.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 89780
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein.
37.4 kDa
75%, by SDS-PAGE with silver staining.
Human
Yes
89780
Wnt-3a
10μg
Unlabeled
Videos