missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGF/PlGF Heterodimer Recombinant Protein

Brand:  R&D Systems™ 297-VP-005

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13462184

  • £298.33 / 5µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: E. coli-derived, Ala27-Arg191 (VEGF) & Ala21-Arg149 (PIGF)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 28 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO

Specifications

Specifications

AAM03108 (VEGF) & AAD30179 (PlGF)
28 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived Human VEGF (Ala27-Arg191) Accession # AAM03108 Human PlGF (Ala21-Arg149) Accession # AAD30179
Unlabeled
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
VEGF/PlGF Heterodimer
5μg
Disulfide-linked heterodimer
Yes
Videos