missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGF-D Recombinant Protein

Brand:  R&D Systems™ 622-VD-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465734

  • £610.53 / 25µg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Phe93-Ser201, with an N-terminal Met and C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 2277
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
13 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2277
VEGF-D
25μg
Unlabeled
Videos