missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGF-C (Cys156Ser) Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Brand:  R&D Systems™ 752-VC-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13466224

  • £298.41 / 25µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human VEGF-C (Cys156Ser) Protein is derived from NS0. The Recombinant Human VEGF-C (Cys156Ser) Protein has been validated for the following applications: Bioactivity, Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
Observed Molecular Wt: 22-24kDa, reducing conditions, Predicted Molecular Wt.: 15.4kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
7424
VEGF-C (Cys156Ser)
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos
Documents