missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human VEGF-C (Cys156Ser) Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 7424 |
Molecular Weight (g/mol) | Observed Molecular Wt: 22-24kDa, reducing conditions, Predicted Molecular Wt.: 15.4kDa |
Name | VEGF-C (Cys156Ser) |
Quantity | 25μg |
Description
The Recombinant Human VEGF-C (Cys156Ser) Protein is derived from NS0. The Recombinant Human VEGF-C (Cys156Ser) Protein has been validated for the following applications: Bioactivity, Binding Activity.Specifications
Unconjugated | |
Observed Molecular Wt: 22-24kDa, reducing conditions, Predicted Molecular Wt.: 15.4kDa | |
25μg | |
Mouse myeloma cell line,NS0-derived human VEGF-C protein Thr103-Arg227 (Cys156Ser),with a C-terminal 10-His tag |
7424 | |
VEGF-C (Cys156Ser) | |
Yes |