missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGF 165 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 293-VE-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13432094

  • £1073.10 / 50µg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human VEGF 165 Protein is derived from Sf 21 (baculovirus). The Recombinant Human VEGF 165 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
19.2 kDa (monomer)
97%, by SDS-PAGE with silver staining.
Human
Disulfide-linked homodimer
Yes
7422
VEGF 165
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents