missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGF 145 (aa 27-171) Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 7626-VE-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436264

  • £357.21 / 10µg

Estimated Shipment date
from Supplier 21-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human VEGF 145 (aa 27-171) Protein is derived from E. coli. The Recombinant Human VEGF 145 (aa 27-171) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
17 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked dimer
Yes
7422
VEGF 145 (aa 27-171)
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents