missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGF 121, aa 207-327 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 4644-VS-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13435014

  • £359.64 / 10µg

Estimated Shipment
17-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human VEGF 121 (aa 207-327) Protein is derived from E. coli. The Recombinant Human VEGF 121 (aa 207-327) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
14 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived human VEGF protein Ala207-Arg327,with an N-terminal Met & Pro208-Arg327
Disulfide-linked homodimer
Yes
7422
VEGF 121, aa 207-327
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Videos
Documents