missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VCAM-1/CD106 Recombinant Protein, (CHO-derived)

Brand:  R&D Systems™ ADP5-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486644

  • £320.77 / 50µg


Estimated Shipment
26-09-2018
Add to basket

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 74.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Entrez Gene ID: 7412
Regulatory Status: RUO

Specifications

Specifications

P19320-1
7412
VCAM-1/CD106
50μg
Unlabeled
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
74.1 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chinese Hamster Ovary cell line, CHO-derived Phe25-Glu698 (Arg322Gly)
Yes
Videos