missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human UBE2I/Ubc9 Recombinant Protein

Brand:  R&D Systems™ E2-645-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464905

  • £253.45 / 100µg


Estimated Shipment
08-01-2019
Add to basket

Description

Description

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 7329
Regulatory Status: RUO

Specifications

Specifications

Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
UBE2I/Ubc9
100μg
Unlabeled
7329
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Yes
Videos