missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TSH alpha/beta Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 4610-TH-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13444984

  • £368.29 / 10µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human TSH alpha/beta Heterodimer Protein is derived from CHO. The Recombinant Human TSH alpha/beta Heterodimer Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
10.2 kDa (TSH alpha), 13.5 kDa (TSH beta)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chinese Hamster Ovary cell line, CHO-derived Human TSH alpha (Ala25-Ser116) Accession # P01215 Human TSH beta (Phe21-Val138) Accession # P01222
Noncovalently-linked heterodimer
Yes
7252
TSH alpha/beta
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Reconstitute at 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Videos