missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TRAIL/TNFSF10 Recombinant Protein, (E. coli-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 375-TEC-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13433564

  • £288.12 / 10µg

Estimated Shipment date
from Supplier 24-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human TRAIL/TNFSF10 (E. coli-expressed) Protein is derived from E. coli. The Recombinant Human TRAIL/TNFSF10 (E. coli-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2 μm filtered solution in Tris, (NH 4 ) 2 SO 4 and ZnSO 4 with BSA as a carrier protein.
19 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Homotrimer
Unlabeled
8743
TRAIL/TNFSF10
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 2 weeks, 2 to 8° C under sterile conditions after reconstitution. 1 month, -20 to -70° C under sterile conditions after reconstitution.
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos
Documents