missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TRAIL/TNFSF10 Recombinant Protein

Brand:  R&D Systems™ 375-TL-010/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463564

  • £348.00 / 10µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Thr95-Gly281, with an N-terminal Met and 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 8743
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
21 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
8743
TRAIL/TNFSF10
10μg
Noncovalently-linked homotrimer and disulfide-linked homodimer
Yes
Videos