missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TL1A/TNFSF15 Recombinant Protein

Brand:  R&D Systems™ 1319-TL-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459133

  • £253.45 / 10µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: E. coli-derived, Leu72-Leu251, with an N-terminal Met
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 9966
Regulatory Status: RUO

Specifications

Specifications

Reconstitute at 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
9966
TL1A/TNFSF15
10μg
Unlabeled
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
21 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
Videos