missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TL1A/TNFSF15 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Gene ID (Entrez) | 9966 |
---|---|
Molecular Weight (g/mol) | 21 kDa (monomer) |
Name | TL1A/TNFSF15 |
Quantity | 10μg |
Source | E. coli-derived human TL1A/TNFSF15 protein Leu72-Leu251,with an N-terminal Met |
Description
The Recombinant Human TL1A/TNFSF15 Protein is derived from E. coli. The Recombinant Human TL1A/TNFSF15 Protein has been validated for the following applications: Bioactivity.Specifications
9966 | |
TL1A/TNFSF15 | |
E. coli-derived human TL1A/TNFSF15 protein Leu72-Leu251,with an N-terminal Met | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
21 kDa (monomer) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |