missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Thrombopoietin/Tpo Recombinant Protein, (NS0-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 288-TPN-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13451984

  • £346.92 / 25µg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Thrombopoietin (NS0-expressed) Protein is derived from NS0. The Recombinant Human Thrombopoietin (NS0-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
35 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
<0.1 EU per 1μg of the protein by the LAL method.
Unlabeled
7066
Thrombopoietin/Tpo
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Yes
Videos
Documents