missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Thrombopoietin/Tpo Recombinant Protein, (NS0-expressed)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

POA

Specifications

Conjugate Unconjugated
Gene ID (Entrez) 7066
Molecular Weight 35 kDa
Name Thrombopoietin/Tpo
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs

Description

Description

The Recombinant Human Thrombopoietin (NS0-expressed) Protein is derived from NS0. The Recombinant Human Thrombopoietin (NS0-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Unconjugated
35 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7066
Thrombopoietin/Tpo
25μg
Mouse myeloma cell line,NS0-derived human Thrombopoietin/Tpo protein Ser22-Gly353