missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Thrombopoietin/Tpo Recombinant Protein, (NS0-expressed)

Brand:  R&D Systems™ 288-TPN-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461984

  • £332.65 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ser22-Gly353
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 35 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 7066
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
35 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
7066
Thrombopoietin/Tpo
25μg
<0.1 EU per 1μg of the protein by the LAL method.
Yes
Videos