missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Thrombopoietin/Tpo Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 288-TP-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441984

  • £867.80 / 25µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Thrombopoietin Protein is derived from Sf 21 (baculovirus). The Recombinant Human Thrombopoietin Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
35 kDa
97%, by SDS-PAGE with silver staining.
Human
Unlabeled
7066
Thrombopoietin/Tpo
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -70° C under sterile conditions after reconstitution.
Yes
Videos