missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Thrombopoietin/Tpo Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 288-TP-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13431984

  • £1026.06 / 25µg

Estimated Shipment date
from Supplier 21-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Thrombopoietin Protein is derived from Sf 21 (baculovirus). The Recombinant Human Thrombopoietin Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
35 kDa
97%, by SDS-PAGE with silver staining.
Human
Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Yes
7066
Thrombopoietin/Tpo
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents