missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human TGF-beta 1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 7040 |
Name | TGF-beta 1 |
Source | Human platelet-derived TGF-beta 1 protein The human platelets used for the isolation of this product were certified by the supplier to be HIV-1 and HBsAg negative at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions. |
Description
The Human TGF-beta 1 Protein is derived from Human Platelets. The Human TGF-beta 1 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
7040 | |
TGF-beta 1 | |
Human platelet-derived TGF-beta 1 protein The human platelets used for the isolation of this product were certified by the supplier to be HIV-1 and HBsAg negative at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions. |