missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TGF-beta 1 Protein

Brand:  R&D Systems™ 100-B-001/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488283

  • £309.00 / 1ยตg


Estimated Shipment
27-07-2018
Add to basket

Description

Description

Source: Human platelet-derived
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 7040
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
TGF-beta 1
1μg
Disulfide-linked homodimer
7040
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
Videos