missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Sonic Hedgehog/Shh (C24II), N-Terminus Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 1845-SH-100

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440114

  • £936.84 / 100µg

Estimated Shipment
10-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus is derived from E. coli. The Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
20 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived human Sonic Hedgehog/Shh protein Cys24-Gly197 (Cys24Ile-Ile),with an N-terminal Met
HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Yes
6469
Sonic Hedgehog/Shh (C24II), N-Terminus
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 100-200 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin, and allow up to 24 hours at 2 to 8° C for complete reconstitution.
Videos
Documents