missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human sFRP-3 Recombinant Protein, C-terminal 10-His tag

Brand:  R&D Systems™ 192-SF-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460244

  • £298.33 / 10µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala33-Asn325
Entrez Gene ID: 2487
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
35 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2487
sFRP-3
10μg
Unlabeled
Videos