missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human sFRP-3 Recombinant Protein

Brand:  R&D Systems™ 7584-SF-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486244

  • £352.00 / 25µg


Estimated Shipment
02-03-2018
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ala33-Asn325
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 33.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 2487
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein.
33.2 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2487
sFRP-3
25μg
Unlabeled
Videos