missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human SCF Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 255-SC-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13431474

  • £730.59 / 50µg

Estimated Shipment date
from Supplier 28-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human SCF Protein is derived from E. coli. The Recombinant Human SCF Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
18.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor
Yes
4254
SCF/c-kit Ligand
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents