missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Relaxin-2 (B-33/A-24) Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 3596-RN-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13443264

  • £245.53 / 25µg

Estimated Shipment date
from Supplier 24-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Relaxin-2 (B-33/A-24) Protein is derived from E. coli. The Recombinant Human Relaxin-2 (B-33/A-24) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
2.7 kDa (A chain), 3.9 kDa (B chain)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked heterodimer
Yes
6019
Relaxin-2 (B-33/A-24)
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents