missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human R-Spondin 4 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 343637 |
Molecular Weight (g/mol) | 24.1 kDa (monomer) |
Name | R-Spondin 4 |
Quantity | 25μg |
Description
The Recombinant Human R-Spondin 4 Protein is derived from CHO. The Recombinant Human R-Spondin 4 Protein has been validated for the following applications: Bioactivity.Specifications
Unconjugated | |
24.1 kDa (monomer) | |
25μg | |
Chinese Hamster Ovary cell line,CHO-derived human R-Spondin 4 protein Met1-Pro234 |
343637 | |
R-Spondin 4 | |
Yes |