missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Protein Fraction I Protein Fraction

Brand:  R&D Systems™ F-375-01M

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13445285

  • £129.60 / 1mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Source: Human cervical epithelial carcinoma cell line, HeLa-derived
Formulation: Supplied as a solution in HEPES.
Regulatory Status: RUO

Specifications

Specifications

Supplied as a solution in HEPES.
1mg
Unlabeled
Protein Fraction I
Human
Videos