missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Protein Fraction I Protein Fraction
POA
Specifications
Conjugate | Unconjugated |
---|---|
Formulation | Supplied as a solution in HEPES. |
Name | Protein Fraction I |
Description
Source: Human cervical epithelial carcinoma cell line, HeLa-derived
Formulation: Supplied as a solution in HEPES.
Regulatory Status: RUO
Specifications
Unconjugated | |
Supplied as a solution in HEPES. | |
Protein Fraction I |