missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Pro EGF aa 21-1023 Recombinant Protein

Brand:  R&D Systems™ 4289-EG-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13464494

  • £266.65 / 25µg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblast cells. Rubin, J.S. et al. (1991) Proc. Natl. Acad. Sci. USA 88:415. The ED 50 for this effect is 1-5 ng/mL.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
112 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
1950
Pro EGF aa 21-1023
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos