missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Placental Lactogen/CSH1 Recombinant Protein

Brand:  R&D Systems™ 5757-PL-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13465534

  • £345.00 / 25µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Val27-Phe217 with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 23.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1442
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
23.1 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
1442
Placental Lactogen/CSH1
25μg
Unlabeled
Videos