missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Pentraxin 3/TSG-14 Recombinant Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
POA
Specifications
Conjugate | Unconjugated |
---|---|
Gene ID (Entrez) | 5806 |
Molecular Weight (g/mol) | 41 kDa (monomer) |
Name | Pentraxin 3/TSG-14 |
Quantity | 25μg |
Description
The Recombinant Human Pentraxin 3/TSG-14 Protein is derived from NS0. The Recombinant Human Pentraxin 3/TSG-14 Protein has been validated for the following applications: Binding Activity.Specifications
Unconjugated | |
41 kDa (monomer) | |
25μg |
5806 | |
Pentraxin 3/TSG-14 | |
Yes |