missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Pentraxin 2/SAP Recombinant Protein

Brand:  R&D Systems™ 1948-SA-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460274

  • £316.81 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, His20-Val223, with a C-terminal 10-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 325
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
25 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
325
Pentraxin 2/SAP
50μg
Noncovalently-linked homopentamer
Yes
Videos