missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human PDGF-AA Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 221-AA-010

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13440824

  • £322.64 / 10µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human PDGF-AA Protein is derived from E. coli. The Recombinant Human PDGF-AA Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
29 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived human PDGF-AA protein Ser87-Thr211,with and without an N-terminal Met
Disulfide-linked homodimer
Yes
5154
PDGF-AA
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Videos
Documents