missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human PDGF-AA Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
POA
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
---|---|
Gene ID (Entrez) | 5154 |
Molecular Weight (g/mol) | M.W. theroretical: 14 kDa (monomer) |
Name | PDGF-AA |
Source | E. coli-derived human PDGF-AA protein Ser87-Thr211,with and without an N-terminal Met |
Description
The Recombinant Human PDGF-AA Protein is derived from E. coli. The Recombinant Human PDGF-AA Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | |
M.W. theroretical: 14 kDa (monomer) | |
E. coli-derived human PDGF-AA protein Ser87-Thr211,with and without an N-terminal Met | |
Disulfide-linked homodimer | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
5154 | |
PDGF-AA | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
PDGF1, PDGF-A, PDGFAA | |
Recombinant | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |