missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human PD-ECGF/Thymidine Phosphorylase Recombinant Protein

Brand:  R&D Systems™ 229-PE-050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460974

  • £700.12 / 50µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala11-Gln482
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 49 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 1890
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
49 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
1890
PD-ECGF/Thymidine Phosphorylase
50μg
Unlabeled
Videos