missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Osteopontin/OPN Recombinant Protein

Brand:  R&D Systems™ 1433-OP-050/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459493

  • £252.13 / 50µg


Estimated Shipment
01-03-2019
Add to basket

Description

Description

Source: Mouse myeloma cell line, NS0-derived, Ile17-Asn300, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 32.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6696
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
32.9 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6696
Osteopontin/OPN
50μg
Unlabeled
Videos