missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human NT-4 Recombinant Protein

Brand:  R&D Systems™ 268-N4-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461734

  • £481.02 / 25µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gly81-Ala210
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 4909
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
14 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
4909
NT-4
25μg
Unlabeled
Videos