missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human NT-3 Recombinant Protein

Brand:  R&D Systems™ 267-N3-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13461714

  • £687.33 / 25µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Tyr139-Thr257 (K196R)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 4908
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
13.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
4908
NT-3
25μg
Unlabeled
Videos