missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Neurturin (Histidine-tagged) Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 387-NE-025

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13443754

  • £194.04 / 25µg

Estimated Shipment date
from Supplier 28-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Neurturin (Histidine-tagged) Protein is derived from E. coli. The Recombinant Human Neurturin (Histidine-tagged) Protein has been validated for the following applications: Bioactivity, Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
14 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked homodimer
Yes
4902
Neurturin (Histidine-tagged)
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos
Documents