missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Neuregulin-1 Isoform SMDF Recombinant Protein

Brand:  R&D Systems™ 378-SM-025/CF

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13463604

  • £364.33 / 25µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Met1-Glu296, Leu117-Glu296 & Ala162-Glu296, each with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 33 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3084
Regulatory Status: RUO

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
33 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
3084
Neuregulin-1 Isoform SMDF
25μg
Unlabeled
Videos